Human Anti-SARS-CoV-2 (S) IgM ELISA Kit
Catalog # 76743-340
Supplier: ANTIBODIES.COM LLC
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Anti-SARS-CoV-2 (S) IgM
- Description:Human anti-SARS-CoV-2 (S) IgM ELISA kit
- Size:96 tests
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0.781 - 50 ng/ml
- Storage temperature:4 °C
- Sensitivity:0.469 ng/ml
- Regulatory status:RUO
- Cat. no.:76743-340
Specifications
About this item
Human Anti-SARS-CoV-2 (S) IgM ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 (S) IgM in serum, plasma, tissue homogenates, and other biological fluids.