Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 500 g Total
Catalog # 470344-944
Supplier: Wards
Specifications
- Description:Zinc casted slotted masses, set of 9 weights, 500 g total
- Environmentally Preferable:
- Cat. no.:470344-944
Specifications
About this item
Set of 9 zinc-casted slotted masses with one hanger each of 50 g, total weight of 500 g. For use in physics experiments and demonstrations.
- Powder-coated, die casted zinc
- 50 g slotted weights with accuracy ±2%
- Complete with hanger
- Great alternative to expensive brass sets
Beautiful die casted zinc 9-piece slotted masses set on durable hanger. Each weight is 50 g, powder-coated, with a ±2% accuracy.