ContactUsLinkComponent
Eisco® Hooked Brass Weights, Set of 9
Eisco® Hooked Brass Weights, Set of 9
Catalog # 470344-474
Supplier:  Wards
undefined
Eisco® Hooked Brass Weights, Set of 9
Catalog # 470344-474
Supplier:  Wards
Supplier Number:  PH0037D
Specifications
  • Description:
    Hooked brass weights, set of 9
  • Environmentally Preferable:
  • Cat. no.:
    470344-474
Specifications
About this item
Set of nine brass hooked weights ranging from 10 to 1000 g, perfect for classroom or lab experiments. Supplied in a wooden block for easy storage.
  • Weights have hooks above and below main body, but can sit flat on a surface due to retractable bottom hook
  • Set comes with wooden storage block
  • Well calibrated, true brass
  • Will last for decades of lab and classroom use
A classic tool used in any lab or classroom. This nine piece brass weight set with wooden block holder will last for decades of use due to its high integrity construction and quality brass material. Set includes: 1000 g, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, and 10 g weights.