Order Entry
Canada
ContactUsLinkComponent
Hot Plate, RC 5
Hot Plate, RC 5
  76795-558
3 year warranty
undefined
Hot Plate, RC 5
  76795-558
 :  F20710610
3 year warranty

 

  • Control type:
    Analog
  • Temperature range:
    310 °C
  • Description:
    RC 5 Hot plate
  • Environmentally Preferable:
  • Model:
    RC 5
  • W×D×H:
    16×27×8.5 cm (6.3×10.6×3.3")
  • Plate dimensions:
    Ø 135 mm
  • Plate material:
    Aluminium alloy
  • Power input/output:
    630 W (power input)
  • Weight:
    1.7 kg (3.7 lbs.)
  • Cat. no.:
    76795-558
  • Electrical:
    115 V, 60 Hz

 

 

The RC 5 is equipped with a 135 mm diameter aluminum alloy top plate ensuring great temperature homogeneity and optimum heat transfer across the entire surface.

  • Easy-to-use, safe and designed to last
  • Two illuminated indicators always inform the operator when the heating is on

You can rely on the RC 5 for everyday laboratory applications such as boiling liquids, basic solutions and media preparation as well as dissolving buffers and reagents. By turning the knob of the RC 5 it is possible to easily adjust the temperature setting up to 310 °C. It accepts up to 15 L flasks.

The extremely intuitive control panel and its inclination have been carefully studied to optimize the user experience for all the lab operators and to offer great visibility. The internal electronic parts are protected by the insulating disk, which, together with the robust die-cast aluminum housing makes the RC 5 a durable and reliable hotplate.

Various safety functions, such as safety circuits and over-temperature protection, protect users in their daily operations. Two illuminated indicators always inform the operator when the heating is on. Furthermore, the icon stays on when the temperature of the plate is above 50 °C even if the instrument is switched off.

 : IP 42 certified.
 : The RC 5 is designed for the most extended lifespan and guaranteed by a 3-year warranty (online product registration required).