Linearity FLQ Cardiac Markers for Roche Systems, AUDIT MircoControls
76778-988
:
- Description:Linearity FLQ Cardiac Markers for Roche Systems
- Size:10x 2 ml
- Cat. no.:76778-988
The Linearity FLQ Cardiac Markers for Roche Systems is an IVD device consisting of 10 levels of liquid material and additives in bovine serum albumin.
- Frozen Liquid Format
The Audit® MicroControlsTM Linearity FLQ Cardiac Markers for Roche Systems is intended to simulate human patient samples for use in determining linearity, calibration verification, and the
verification of reportable range for the following analytes: CKMB, Myoglobin, TNI, TNT.
: For In Vitro Diagnostic use only.