ContactUsLinkComponent
Linearity FLQ Fertility for Siemens Atellica/Centaur, AUDIT MircoControls
Linearity FLQ Fertility for Siemens Atellica/Centaur, AUDIT MircoControls
Catalog # 76778-984
CAS Number:  
undefined
Linearity FLQ Fertility for Siemens Atellica/Centaur, AUDIT MircoControls
Catalog # 76778-984
Supplier Number:  K818M-5
CAS Number:  
Specifications
  • Description:
    Linearity FLQ Fertility for Siemens Atellica/Centaur
  • Size:
    10x 2 ml
  • Cat. no.:
    76778-984
Specifications
About this item
The Audit® MicroControlsTM Linearity FLQ Fertility for Siemens Atellica/Centaur is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: AFP, DHEA-S, Estradiol, Progesterone, FSH, HCG, LH, Prolactin, Testosterone.
  • Frozen Liquid Format
Consists of 10 levels of liquid material and additives in bovine serum albumin.
Caution: For In Vitro Diagnostic use only.