Anti-Telomerase Binding Protein, P23 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
76101-210
: Bioss
- Antibody type:Primary
- Antigen name:Telomerase Binding Protein, P23
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µl
- Epitope:31-130/160
- Form:Liquid
- Gene ID:10728
- Antigen synonyms:PTGES3|P23|Prostaglandin E synthase 3|Cytosolic prostaglandin E2 synthase|Telomerase-binding protein p23|TEBP_HUMAN.|cPGES|TEBP|Progesterone receptor complex p23|Telomerase Binding Protein???P23|Hsp90 co-chaperone
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7,4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular weight:18 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze–thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Telomerase Binding Protein, P23
- Tested applications:ICC
- Purification:Purified by Protein A
- Cat. no.:76101-210
- Packaging:Vial
Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Belongs to the p23/wos2 family.
Type: Primary
Antigen: Telomerase Binding Protein, P23
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 31-130/160
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat