To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:PI14
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Epitope:151-250/405
- Form:Liquid
- Gene ID:5276
- Antigen synonyms:Serine or cysteine proteinase inhibitor clade I neuroserpin member 2|Pancpin|Myoepithelium derived serine protease inhibitor|MEPI|TSA2004|Serpin peptidase inhibitor clade I pancpin member 2|Serpin I2|PI14|Peptidase inhibitor 14|SERPINI2|PI 14|Pancreas specic protein TSA2004|Serine or cysteine proteinase inhibitor clade I pancpin member 2|Protease inhibitor 14|SPI2_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SERPINI2/PI14
- Purification:Purified by Protein A
- Cat. no.:76110-718
- Packaging:Vial
SERPINI2 is a member of the serine protease inhibitor (serpin) superfamily made up of proteins which play central roles in the regulation of a wide variety of physiological processes, including coagulation, fibrinolysis, development, malignancy and inflammation. It may have a role in a growth-control, possibly growth-suppressing pathway and, when impaired, may be involved in pancreatic carcinogenesis. The protein is a member of the plasminogen activator inhibitor-1 family, a subset of the serpin superfamily whose members act as tissue-specific tPA inhibitors. Two alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene.
Type: Primary
Antigen: PI14
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/405
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat