To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:NFAT5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Epitope:221-320/1531
- Form:Liquid
- Gene ID:10725
- Antigen synonyms:TonE-binding protein |NF-AT5|NFATL1|NF AT5|NFAT 5|Tonicity-responsive enhancer-binding protein|NFAT5|T-cell transcription factor NFAT5|T cell transcription factor NFAT5|Nuclear factor of activated T cells 5 tonicity responsive|Nuclear factor of activated T-cells 5|TonEBP|NFATZ|KIAA0827|TonE binding protein|Nuclear factor of activated T cells 5|NFAT5_HUMAN|Osmotic response element binding protein|Glutamine rich protein H65|NFAT L1|Nuclear factor of activated T cells|OREBP|T cell transcription factor NFAT 5|NFAT like protein 1|NFATL 1|Tonicity responsive enhancer binding protein
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human NFAT5
- Purification:Purified by Protein A
- Cat. no.:76110-688
- Packaging:Vial
Specifications
About this item
The product of this gene is a member of the nuclear factors of activated T cells family of transcription factors. Proteins belonging to this family play a central role in inducible gene transcription during the immune response. This protein regulates gene expression induced by osmotic stress in mammalian cells. Unlike monomeric members of this protein family, this protein exists as a homodimer and forms stable dimers with DNA elements. Multiple transcript variants encoding different isoforms have been found for this gene.
Type: Primary
Antigen: NFAT5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 221-320/1531
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat