To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B-Raf Ser365
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Size:100 µl
- Form:Liquid
- Gene ID:673
- Antigen synonyms:NS7|p94|Serine/threonine-protein kinase B-raf|Proto-oncogene B-Raf|BRAF|RAFB1|v-Raf murine sarcoma viral oncogene homolog B1|BRAF1|B-RAF1
- Modification:Phosphorylation (Ser365)
- Storage buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular weight:84 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human B Raf around the phosphorylation site of Ser365
- Tested applications:ICC
- Purification:Purified by Protein A
- Cat. no.:76083-252
- Packaging:Vial
Specifications
About this item
Protein kinase involved in the transduction of mitogenic signals from the cell membrane to the nucleus. May play a role in the postsynaptic responses of hippocampal neuron. Phosphorylates MAP2K1, and thereby contributes to the MAP kinase signal transduction pathway.
Type: Primary
Antigen: B-Raf Ser365
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat