Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # 76101-730
Supplier: Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:duck hepatitis A virus 1 Genome polyprotein
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Other,Virus
- Western blot:Yes
- Size:100 µl
- Epitope:851-950/2249
- Form:Liquid
- Antigen synonyms:DHAV|polyprotein DHAV|polyprotein [duck hepatitis A virus]
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular weight:252 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:MAP peptide derived from polyprotein [duck hepatitis A virus] FC64/CHN/2009
- Tested applications:ICC
- Purification:Purified by Protein A
- Cat. no.:76101-730
- Packaging:Vial
Specifications
About this item
Duck hepatitis A virus genome polyprotein.
Type: Primary
Antigen: duck hepatitis A virus 1 Genome polyprotein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 851-950/2249
Host: Rabbit
Isotype: IgG
Reactivity: Virus; Others