Anti-LDOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # 76118-342
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:LDOC1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µl
- Epitope:65-146/146
- Form:Liquid
- Gene ID:23641
- Antigen synonyms:Leucine zipper protein down-regulated in cancer cells|Protein LDOC1|LDOC1|Mar7|BCUR1|Mart7
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular weight:17 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human LDOC1
- Tested applications:ICC
- Purification:Purified by Protein A
- Cat. no.:76118-342
- Packaging:Vial
Specifications
About this item
May have an important role in the development and/or progression of some cancers.
Type: Primary
Antigen: LDOC1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 65-146/146
Host: Rabbit
Isotype: IgG
Reactivity: