To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CLLD8
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Epitope:21-120/719
- Form:Liquid
- Gene ID:83852
- Antigen synonyms:Chronic lymphocytic leukemia deletion region gene 8 protein|SET domain burcated 2|Chronic lymphocytic leukemia deletion region 8|SEB2|H3-K9-HMTase|Chronic lymphocytic leukemia deletion region gene 8|Setdb2|KMT1F|Clld8|Probable histone-lysine N-methyltransferase H3 lysine-9 specic|SETB2_HUMAN|Histone-lysine N-methyltransferase SETDB2|CLLL8|Chromosome 13 open reading frame 4|SET domain protein, burcated, 2|Histone H3-K9 methyltransferase|Lysine N-methyltransferase 1F|Gm293|C13orf4
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular weight:79 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CLLD8/SETDB2
- Tested applications:ICC
- Purification:Purified by Protein A
- Cat. no.:76117-248
- Packaging:Vial
Specifications
About this item
Histone methyltransferase involved in left-right axis specification in early development and mitosis. Specifically trimethylates 'Lys-9' of histone H3 (H3K9me3). H3K9me3 is a specific tag for epigenetic transcriptional repression that recruits HP1 (CBX1, CBX3 and/or CBX5) proteins to methylated histones. Contributes to H3K9me3 in both the interspersed repetitive elements and centromere-associated repeats. Plays a role in chromosome condensation and segregation during mitosis.
Type: Primary
Antigen: CLLD8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/719
Host: Rabbit
Isotype: IgG
Reactivity: Rat