Anti-SERTAD2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
76117-210
: Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:SERTAD2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Environmentally Preferable:
- Epitope:111-210/314
- Form:Liquid
- Gene ID:9792
- Antigen synonyms:SRTD2_HUMAN|SERTA domain-containing protein 2|MGC126690|TRIP-Br2|Transcriptional regulator interacting with the PHS-bromodomain 2|Sei-2|KIAA0127|Sei2|Transcriptional regulator interacting with the PHD-bromodomain 2|Transcriptional regulator interacting with the PHD bromodomain 2|SERTA domain containing 2|TRIP Br2|Sei 2|MGC126688
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular weight:34 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SERTAD2
- Tested applications:ICC
- Purification:Purified by Protein A
- Cat. no.:76117-210
- Packaging:Vial
SERTA domain containing 2 (SERTAD2) acts at E2F-responsive promoters to integrate signals provided by PHD- and/or bromodomain-containing transcription factors.
Type: Primary
Antigen: SERTAD2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 111-210/314
Host: Rabbit
Isotype: IgG
Reactivity: Human