Specifications
- Description:Purilux vent 2×2' LED light panel
- Cat. no.:76716-790
Specifications
About this item
Purilux Vent 2×2' LED Light Panel
- Easy to install; fits like a 2×2' troffer/panel
- Vibration- and impact-resistant
- Quiet performance
- Emergency battery backup option available
- Instant on/off
Originally developed in partnership with epidemiologists at Boston Children’s Hospital, the Purilux silently produces the purest, safest air possible, while also providing a modern, energy efficient lighting solution. The air in buildings equipped with the Purilux is free from airborne pathogens, viruses, mold, environmental pollution, allergens, and other irritants for maximum safety and wellness. Built-in ULPA and carbon filters team up with a medical grade, centrifugal fan and a UVC/UVA sanitizing chamber to purify the air, eliminating viruses and other airborne risks, including TVOCs, without the use of any chemicals, ions, or ozone.
Studies show that clean, healthy indoor air significantly lessens the transmission of disease, boosts focus and productivity for workers, improves cognitive function, leads to less absenteeism due to illness, reduces the impact of asthma and allergies, supports deep, restful sleep, and lowers the risk of respiratory illness.