ContactUsLinkComponent
CE Duo® CleanTech® Non Sterile Mops with Bucket Liner
CE Duo® CleanTech® Non Sterile Mops with Bucket Liner
Catalog # 76635-846
undefined
CE Duo® CleanTech® Non Sterile Mops with Bucket Liner
Catalog # 76635-846
Supplier Number:  172474
Specifications
  • Color:
    Blue
  • Description:
    CE Duo® Cleantech® Nonsterile cleanroom foam mop
  • Cat. no.:
    76635-846
  • Mop Head Width:
    19.7"
Specifications
About this item
This mop is now available in a conveinent six pack with a bucket liner. Cutting down not only on the time it takes to prepare the mops but also significantly reducing the amount of packaging waste.
  • Microfiber, overstitched cleanroom foam for cleaning/disinfecting floors, walls, and ceilings
  • Adhesive-free lamination techniqe prevents additional contamination
  • Excellent microfiber cleaning results have been independently proven
  • Reduce packaging waste
  • Reduce amount of time needed to pre-prepare the mops
CE Duo® Cleantech® nonsterile mop is a single-use, double-sided microfiber cleanroom foam mop for cleaning and disinfecting floors, walls, and ceilings. The CE Duo® CleanTech® mop is packaged 6 mops per pack, single bagged with bucket liner.
Packaging: Mops are packaged 6 per pack, 8 packs per case.