Eisco Weather Station Instrument Box
Catalog # 470350-884
Supplier: Wards
Specifications
- Description:Weather station instrument box
- Cat. no.:470350-884
Specifications
About this item
GLOBE®-certified ventilated lockable shelter for protecting valuable weather instruments.
- GLOBE®-certified: meets the stringent specifications presented in the GLOBE Teacher's Guide
- Weather-resistant composite construction
- Louvered sides allow ambient outdoor readings while keeping out damaging wind, rain, and other elements
- Lockable hinged door allows easy access and prevents tampering
- Measures 23H×11W×9"D
- Aligned with GLOBE protocols
A louvered box of painted composite wood provides a ventilated shelter for instruments that need to be protected from the elements, yet still need to be outdoors to take measurements. Comes with two keys for locking the hinged access door to prevent tampering.
Certifications: GLOBE-Certified.