Thermo Scientific® TSG Pharmacy Refrigerators
Supplier: Thermo Fisher Scientific
Certificates
About this item
Thermo Scientific TSG Series pharmacy refrigerators are specialized for clinical and pharmaceutical applications, offering quiet, intuitive operation with excellent temperature performance and an outstanding warranty. Validated NSF 456 compliant performance means TSG Series pharmacy refrigerators deliver exceptional security and temperature stability for vaccines, pharmaceuticals and other precious cargo.
- Specialized for clinical and pharmaceutical applications
- Validated NSF 456 compliance
- Independent temperature display probe
- Quiet operation, less than 53 dB
- Heat-free defrost, ENERGY STAR certified
- Double pane glass door, keyed lock
- Hi/Lo temperature, door ajar, and power failure alarms and remote alarm contacts
- 4 years parts and labor and 1 year compressor warranty
All TSG Series refrigerators are built with sustainability in mind to minimize power consumption without sacrificing performance at a zero waste to landfill facility.
Certifications: cULus, ENERGYSTAR.
Ordering information: Accessories available include chart recorders, independent temperature monitors, surge suppressors and more.
Specifications
- Catalog No:
- CA76764-966
- CA76764-968
- CA76764-982
- CA76764-954
- CA76764-984
- CA76764-956
- CA76764-944
- CA76764-942
- Environmentally Preferable:
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health, Low Manufacturing Impact, Energy Efficient, Product Transparency and Disclosure
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health, Low Manufacturing Impact, Energy Efficient, Product Transparency and Disclosure
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health, Low Manufacturing Impact, Energy Efficient, Product Transparency and Disclosure
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health, Low Manufacturing Impact, Energy Efficient, Product Transparency and Disclosure
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health, Low Manufacturing Impact, Energy Efficient, Product Transparency and Disclosure
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health, Low Manufacturing Impact, Energy Efficient, Product Transparency and Disclosure
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health, Low Manufacturing Impact, Energy Efficient, Product Transparency and Disclosure
- Sustainable Packaging, Sustainable Materials, Safer Human and Environmental Health, Low Manufacturing Impact, Energy Efficient, Product Transparency and Disclosure
- Cat. No.:
- CA76764-966
- CA76764-968
- CA76764-982
- CA76764-954
- CA76764-984
- CA76764-956
- CA76764-944
- CA76764-942
- Defrost:
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Automatic
- Material:
- Painted steel
- Painted steel
- Painted steel
- Painted steel
- Painted steel
- Painted steel
- Painted steel
- Painted steel
- Refrigerant:
- R290
- R290
- R290
- R290
- R290
- R290
- R290
- R290
- Plug:
- NEMA 6-15
- NEMA 6-15
- NEMA 6-15P
- NEMA 6-15P
- NEMA 6-15
- NEMA 6-15
- NEMA 6-15
- NEMA 6-15P
- Environmental certification:
- ENERGY STAR, Significant New Alternatives Policy (SNAP)
- ENERGY STAR, Significant New Alternatives Policy (SNAP)
- ENERGY STAR, Significant New Alternatives Policy (SNAP)
- ENERGY STAR, Significant New Alternatives Policy (SNAP)
- ENERGY STAR, Significant New Alternatives Policy (SNAP)
- ENERGY STAR, Significant New Alternatives Policy (SNAP)
- ENERGY STAR, Significant New Alternatives Policy (SNAP)
- ENERGY STAR, Significant New Alternatives Policy (SNAP)