Eisco Glass Desiccators with Knob Cover and Porcelain Plate
Supplier: Wards
Certificates
About this item
Glass desiccator with knob lid and unglazed procelain plate with five 1" holes. Made from high quality, heavy duty, 3.3 borosilicate glass.
- Holds a full vacuum (740 mm Hg 20 cm) at room temperature for 24 hours - will remain airtight even if not under vacuum
- Chamber and lid are made of high quality semi-frosted borosilicate 3.3 glass
- Features a knob cover
- Unglazed, porcelain sieve plate
- Five perforation holes each measure 1" in diameter
Low temperature gradient- can withstand high temperatures and thermal shocks. Heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Delivery: Supplied with knob lid and unglazed porcelain plate.
Customers Who Bought This Also Bought