- Calibration:pH: automatic one, two, or three points with automatic recognition of five standard buffers (pH 4.01, 6.86, 7.01, 9.18, 10.01) or one custom buffer
ORP: automatic at one custom point (relative mV)
Conductivity: automatic single point, with six standard solutions (84 µS/cm, 1413 µS/cm, 5.00 mS/cm, 12.88 mS/cm, 80.0 mS/cm, 111.8 mS/cm) or custom point
TDS: based on conductivity or salinity calibration
Dissolved oxygen: automatic one or two points at 0 and 100% or one custom point
Salinity: based on conductivity calibration
Resistivity: based on conductivity or salinity calibration - Description:Multiparameter pH/EC/DO bluetooth meter
- DO range:0.0 - 500.0%, 0.00 - 50.00 ppm (mg/L)
- DO resolution:0.1%, 0.01 ppm (mg/L)
- Conductivity accuracy:±1% of reading or ±1 µS/cm whichever is greater
- Conductivity range:0 - 200 mS/cm (absolute EC up to 400 mS/cm)
- Conductivity resolution:Manual: 1 µS/cm, 0.001 mS/cm, 0.01 mS/cm, 0.1 mS/cm, 1 mS/cm Automatic: 1 µS/cm from 0 - 9999 µS/cm, 0.01 mS/cm from 10.00 - 99.99 mS/cm, 0.1 mS/cm from 100.0 - 400.0 mS/cm Automatic (mS/cm): 0.001 mS/cm from 0.000 - 9.999 mS/cm, 0.01 mS/cm from 10.00 - 99.99 mS/cm, 0.1 mS/cm from 100.0 - 400.0 mS/cm
- DO accuracy:0.1%, 0.01 ppm (mg/L)
- Resistivity range:0 - 999999 Ω•cm, 0 - 1000.0 kΩ•cm, 0 - 1.0000 MΩ•cm
- Salinity range:0.00 - 70.00 PSU
- Salinity resolution:0.01 PSU
- TDS accuracy:±1% of reading or ±1 ppm (mg/L) whichever is greater
- TDS range:0 - 400000 ppm (mg/L) (the maximum value depends on the TDS factor)
- TDS resolution:Manual: 1 ppm (mg/L), 0.001 ppt (g/L), 0.01 ppt (g/L), 0.1 ppt (g/L), 1 ppt (g/L)
Automatic: 1 ppm (mg/L) from 0 - 9999 ppm (mg/L), 0.01 ppt (g/L) from 10.00 - 99.99 ppt (g/L), 0.1 ppt (g/L) from 100.0 - 400.0 ppt (g/L)
Automatic ppt (g/L): 0.001 ppt (g/L) from 0.000 - 9.999 ppt (g/L), 0.01 ppt (g/L) from 10.00 - 99.99 ppt (g/L), 0.1 ppt (g/L) from 100.0 - 400.0 ppt (g/L) - mV accuracy:0.1 mV (ORP)
- mV range:±2000.0 mV (ORP)
- mV resolution:0.1 mV (ORP)
- pH accuracy:±0.02 pH
- pH range:0.00 - 14.00 pH
- pH resolution:0.01 pH
- Cat. no.:CA470349-832
Functional and accurate, this meter is capable of testing 12 different water quality parameters using pH, EC, and optical DO sensors. Transfer data to a smart device for review or sharing with the integrated Bluetooth connection and Hanna Lab App.
- Waterproof (meter rated IP 67, probe rated IP 68)
- Automatic interval logging of up to 45,000 samples or log-on-demand
- Perfect for environmental and industrial professional
- Measure up to 12 parameters
- Don't lose important data with automatic interval logging of up to 45,000 samples or choose to log-on-demand to store measurement data for key points
- For better traceability, calibration information is recorded along with a time and date stamp and stored with logged data
Built durable - Designed to withstand the knocks, drops, and spills of real life, the IP 67 body ensures top performance in any environment. These meters are protected against dust and water intrusion from any direction.
Multiparameter probe - Sensor replacement is quick and easy with field replaceable, screw type connectors and are color-coded for easy identification.
Measurement and graphing - Select the measurement units to be displayed on your smart device. For trend analysis, take a look at the graph of your logged data. Comprehensive GLP data can be reviewed for all parameters when the logged data is downloaded to your smart device.
Sort and share - Group your data by time or notation. Email it for storage or share it with friends or colleagues.