Orders LinkContactUsLinkComponent
phi29-XT RCA Kit
phi29-XT RCA Kit
New Product
phi29-XT RCA Kit
New Product
Certificates
phi29-XT RCA Kit
phi29-XT RCA Kit
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The phi29-XT RCA DNA amplification kit includes everything needed for sensitive and robust rolling circle amplification (RCA), an isothermal amplification approach to continuously amplify circular DNA by generating long, repetitive copies of the circular sequence.
  • High sensitivity: amplify from as little as 1 fg of input plasmid DNA
  • Powered by phi29-XT DNA Polymerase, an engineered polymerase
  • Simple workflow produces high yield in a short reaction time
  • Includes exonuclease-resistant random primers and dNTPs
  • Flexible protocol allows for different types of input material
RCA enables several downstream applications, including DNA sequencing, cell-free protein expression, viral DNA enrichment, plasmid amplification, and padlock probe-based assays. The kit features phi29-XT DNA polymerase, an engineered polymerase with improved thermostability, sensitivity, and capable of generating higher yield in a shorter reaction time than wild-type phi29 DNA polymerase.
Also included are exonuclease-resistant random primers (containing phosphorothioate bonds) to universally amplify any circular DNA sequence and dNTPs. Input material can be a purified plasmid, liquid media culture, a cultured colony from an agar plate, or a glycerol stock.
Product Family Options
Product Information
  • No. of reactions
    Availability
    Price
  • CA76717-052
    phi29-XT RCA DNA Amplification kit
    phi29-XT RCA DNA Amplification kit
    500
    Log in for availability
    Log in for availability
    Each
    Each - 2 200,00 $
    2 200,00 $
    Each
       2 200,00 $
    Supplier Number: E1603L
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • CA76717-046
    phi29-XT RCA DNA Amplification kit
    phi29-XT RCA DNA Amplification kit
    100
    Log in for availability
    Log in for availability
    Each
    Each - 518,00 $
    518,00 $
    Each
       518,00 $
    Minimum order quantity is 1
    Supplier Number: E1603S
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)