Human L-Fucose Kinase(FUK) ELISA Kit
76699-980
:
- Assay Type:Sandwich
- Host:Human
- Target protein:L-fucose kinase
- Description:Human L-fucose kinase(FUK) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:25 - 1600 pg/ml
- Storage temperature:2…8 °C
- Application:Metabolism
- Sample volume:50 - 100 µl
- Sensitivity:6 pg/ml
- Regulatory status:RUO
- Cat. no.:76699-980
- Detection wavelength: 450 nm