Human Dolichyl-Diphosphooligosaccharide-Protein Glycosyltransferase(DDOST,OST) ELISA Kit
Catalog # 76713-476
Supplier: AFG Bioscience
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:Dolichyl-diphosphooligosaccharide-protein glycosyltransferase
- Description:Human dolichyl-diphosphooligosaccharide-protein glycosyltransferase(DDOST,OST) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.625 - 20 ng/ml
- Storage temperature:2…8 °C
- Application:Signal Transduction
- Sample volume:50 - 100 µl
- Sensitivity:0.078 ng/ml
- Regulatory status:RUO
- Cat. no.:76713-476
Specifications
About this item
- Detection wavelength: 450 nm