Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Sialic Acid(SA) ELISA Kit

Human Sialic Acid(SA) ELISA Kit

Human Sialic acid(SA) ELISA Kit, 96 wells
Catalog # 76702-390
Supplier:   AFG Bioscience
Supplier #:  EK711403
Human Sialic Acid(SA) ELISA Kit
Catalog # 76702-390
Supplier:  AFG Bioscience
CAS Number:  
Human Sialic Acid(SA) ELISA Kit
Human Sialic Acid(SA) ELISA Kit

Human Sialic acid(SA) ELISA Kit, 96 wells

CAS Number:  
Price
$678.08
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
  • Detection wavelength: 450 nm
Specifications

Specifications

  • Description:
    Human Sialic acid(SA) ELISA Kit
  • Size:
    96 wells
  • Sample type:
    Serum, plasma, urine, tissue homogenates, cell culture supernates
  • Sample volume:
    50 - 100 µl
  • Detection range:
    16 - 980 nmol/ml
  • Storage temperature:
    2…8 °C
  • Host:
    Human
  • Shelf life:
    6 months (4 °C); 12 months (–20 °C)
  • Assay Type:
    Sandwich
  • Time to Results:
    1 - 5 h
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76702-390
  • Sensitivity:
    4.5 nmol/ml
  • Target protein:
    Sialic acid
  • Regulatory status:
    RUO

Specifications