Human Immunoglobulin G(IgG) ELISA Kit
76711-974
:
- Assay Type:Sandwich
- Host:Human
- Target protein:Immunoglobulin G
- Description:Human Immunoglobulin G(IgG) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:3 - 70 μg/ml
- Storage temperature:2…8 °C
- Application:Immunology
- Sample volume:50 - 100 µl
- Sensitivity:1 μg/ml
- Regulatory status:RUO
- Cat. no.:76711-974
- Detection wavelength: 450 nm