Human Aniti-Phosphatidyl Serine Antibody(APSA) ELISA Kit
76709-804
:
- Assay Type:Sandwich
- Host:Human
- Target protein:Anti-phosphatidyl serine antibody
- Description:Human Anti-phosphatidyl serine antibody(APSA) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Qualitative
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Storage temperature:2…8 °C
- Application:Immunology
- Sample volume:50 - 100 µl
- Sensitivity:Positive/Negative
- Regulatory status:RUO
- Cat. no.:76709-804
- Detection wavelength: 450 nm