Rat Fibroblast Growth Factor-3 (FGF-3) ELISA Kit
Catalog # 76702-256
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:Fibroblast growth factor-3
- Description:Rat fibroblast growth factor-3 (FGF-3) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Application:Cardiovascular, signal transduction
- Sample volume:50 - 100 µl
- Regulatory status:RUO
- Cat. no.:76702-256
Specifications
About this item
- Detection wavelength: 450 nm