Rat Interleukin-3 Receptor Subunit alpha (IL3RA) ELISA Kit
76715-110
:
- Assay Type:Sandwich
- Host:Rat
- Target protein:Interleukin-3 receptor subunit alpha
- Description:Rat Interleukin-3 receptor subunit alpha(IL3RA) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Application:Immunology
- Sample volume:50 - 100 µl
- Regulatory status:RUO
- Cat. no.:76715-110
- Detection wavelength: 450 nm