Human Follicular Dendritic Cell Secreted Peptide (C4orf7) ELISA Kit
Catalog # 76708-636
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:Follicular dendritic cell secreted peptide
- Description:Human Follicular dendritic cell secreted peptide(C4orf7) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Sample volume:50 - 100 µl
- Sensitivity:1 5 h
- Regulatory status:RUO
- Cat. no.:76708-636
Specifications
About this item
- Detection wavelength: 450 nm