Human Baculoviral IAP Repeat-containing Protein 2 (BIRC2) ELISA Kit
76710-956
:
- Assay Type:Sandwich
- Host:Human
- Target protein:Baculoviral IAP repeat-containing protein 2
- Description:Human Baculoviral IAP repeat-containing protein 2(BIRC2) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Application:Cell biology
- Sample volume:50 - 100 µl
- Sensitivity:1 5 h
- Regulatory status:RUO
- Cat. no.:76710-956
- Detection wavelength: 450 nm