Human Gamma-secretase Subunit APH-1A (APH1A) ELISA Kit
76713-372
:
- Assay Type:Sandwich
- Host:Human
- Target protein:Gamma-secretase subunit APH-1A
- Description:Human Gamma-secretase subunit APH-1A(APH1A) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Detection range:62.5 - 2000 pg/ml
- Storage temperature:2…8 °C
- Sample volume:50 - 100 µl
- Sensitivity:7 pg/ml
- Regulatory status:RUO
- Cat. no.:76713-372
- Detection wavelength: 450 nm