Human Angiopoietin-1 (ANGPT1) ELISA Kit
Catalog # 76713-860
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:Angiopoietin-1
- Description:Human Angiopoietin-1(ANGPT1) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Detection range:3.75 - 120 ng/ml
- Storage temperature:2…8 °C
- Application:Cardiovascular
- Sample volume:50 - 100 µl
- Sensitivity:0.091 ng/ml
- Regulatory status:RUO
- Cat. no.:76713-860
Specifications
About this item
- Detection wavelength: 450 nm