Human A Disintegrin and MetalloProteinase with Thrombospondin Motifs 9 (ADAMTS9)
Catalog # 76702-186
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:A disintegrin and metalloproteinase with thrombospondin motifs 9
- Description:Human A disintegrin and metalloproteinase with thrombospondin motifs 9(ADAMTS9)
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Detection range:2 - 64 ng/ml
- Storage temperature:2…8 °C
- Application:Cancer, cardiovascular
- Sample volume:50 - 100 µl
- Sensitivity:0.195 ng/ml
- Regulatory status:RUO
- Cat. no.:76702-186
Specifications
About this item
- Detection wavelength: 450 nm