Human β-actin ELISA Kit
76713-322
:
- Assay Type:Sandwich
- Host:Human
- Target protein:Β-actin
- Description:Human β-actin ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Detection range:0.625 - 20 ng/ml
- Storage temperature:2…8 °C
- Application:Signal transduction
- Sample volume:50 - 100 µl
- Sensitivity:0.078 ng/ml
- Regulatory status:RUO
- Cat. no.:76713-322
- Detection wavelength: 450 nm