Human Transforming Growth Factor, beta 2 (TGFB2) AutoAntibody ELISA Kit
Catalog # 76715-072
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:Transforming growth factor, beta 2
- Description:Human transforming growth factor, beta 2 (TGFB2) autoantibody ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Application:Signal transduction
- Sample volume:50 - 100 µl
- Regulatory status:RUO
- Cat. no.:76715-072
Specifications
About this item
- Detection wavelength: 450 nm