Human Anti-toxoplasmosis (TOXO) Antibody (IgM) ELISA Kit
Catalog # 76714-100
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Capture
- Host:Human
- Target protein:Anti-toxoplasmosis
- Description:Human anti-toxoplasmosis (TOXO) antibody (IgM) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Qualitative
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Sample volume:50 - 100 µl
- Regulatory status:RUO
- Cat. no.:76714-100
Specifications
About this item
- Detection wavelength: 450 nm