Human Anti Respiratory Syncytial Virus (RSV) Antibody (IgG) ELISA Kit
76716-022
:
- Assay Type:Sandwich
- Host:Human
- Target protein:Anti respiratory syncytial virus
- Description:Human anti respiratory syncytial virus (RSV) antibody (IgG) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Qualitative
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Application:Microbiology
- Sample volume:50 - 100 µl
- Regulatory status:RUO
- Cat. no.:76716-022
- Detection wavelength: 450 nm