Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Carboxyhemoglobin, HbCO ELISA Kit

Human Carboxyhemoglobin, HbCO ELISA Kit

Human carboxyhemoglobin,HbCO ELISA Kit, 96 wells
Catalog # 76706-840
Supplier:   AFG Bioscience
Supplier #:  EK714418
Human Carboxyhemoglobin, HbCO ELISA Kit
Catalog # 76706-840
Supplier:  AFG Bioscience
CAS Number:  
Human Carboxyhemoglobin, HbCO ELISA Kit
Human Carboxyhemoglobin, HbCO ELISA Kit

Human carboxyhemoglobin,HbCO ELISA Kit, 96 wells

CAS Number:  
Price
$678.08
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
  • Detection wavelength: 450 nm
Specifications

Specifications

  • Description:
    Human carboxyhemoglobin,HbCO ELISA Kit
  • Size:
    96 wells
  • Sample type:
    Serum, plasma, urine, tissue homogenates, cell culture supernates
  • Sample volume:
    50 - 100 µl
  • Detection range:
    62.5 - 2000 ng/ml
  • Storage temperature:
    2…8 °C
  • Host:
    Human
  • Shelf life:
    6 months (4 °C); 12 months (−20 °C)
  • Assay Type:
    Sandwich
  • Time to Results:
    1 - 5 h
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76706-840
  • Sensitivity:
    0.195 ng/ml
  • Target protein:
    Carboxyhemoglobin,HbCO
  • Regulatory status:
    RUO
  • Application:
    Cardiovascular

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.