Order Entry
Canada
ContactUsLinkComponent
Human Voltage-gated Potassium Channel Antibody (VGKC-ab) ELISA Kit
Human Voltage-gated Potassium Channel Antibody (VGKC-ab) ELISA Kit
Catalog # 76709-610
Supplier:  AFG Bioscience
CAS Number:  
undefined
Human Voltage-gated Potassium Channel Antibody (VGKC-ab) ELISA Kit
Catalog # 76709-610
Supplier:  AFG Bioscience
Supplier Number:  EK714293
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Human
  • Target protein:
    Voltage-gated potassium channel antibody
  • Description:
    Human voltage-gated potassium channel antibody (VGKC-ab) ELISA Kit
  • Size:
    96 wells
  • Sample type:
    Serum, plasma, urine, tissue homogenates, cell culture supernates
  • Time to Results:
    1 - 5 h
  • Shelf life:
    6 months (4 °C); 12 months (−20 °C)
  • Storage temperature:
    2…8 °C
  • Sample volume:
    50 - 100 µl
  • Regulatory status:
    RUO
  • Cat. no.:
    76709-610

Specifications

About this item

  • Detection wavelength: 450 nm