Human Receptor Interacting Serine, Threonine Protein Kinase 3 (RIPK3) ELISA Kit
Catalog # 76707-244
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:Receptor-interacting serine,threonine-protein kinase 3
- Description:Human Receptor-interacting serine,threonine-protein kinase 3(RIPK3) ELISA Kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Detection range:15.6 - 1000 pg/ml
- Storage temperature:2…8 °C
- Application:Cancer, signal transduction
- Sample volume:50 - 100 µl
- Sensitivity:2 pg/ml
- Regulatory status:RUO
- Cat. no.:76707-244
Specifications
About this item
- Detection wavelength: 450 nm