Sheep Calprotectin (CALP) ELISA Kit
Catalog # 76700-942
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Sheep
- Target protein:Calprotectin
- Description:Sheep Calprotectin (CALP) ELISA kit
- Size:96 wells
- Sample type:Serum, plasma, urine,tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:1 - 80 ng/ml
- Storage temperature:2…8 °C
- Sample volume:50 - 100 μl
- Sensitivity:0.1 ng/ml
- Regulatory status:RUO
- Cat. no.:76700-942
Specifications
About this item
- Detection wavelength: 450 nm