Orders LinkContactUsLinkComponent
Premium Beaker Set
Premium Beaker Set
50 / 100 / 250 ml, Premium beaker set
Catalog # 470345-912
Supplier:   Wards
Supplier #:  CH2019005
Premium Beaker Set
Catalog # 470345-912
Supplier:  Wards
Premium Beaker Set
Premium Beaker Set
50 / 100 / 250 ml, Premium beaker set
Price
$20.30
Log in for availability
Each
Product Details & Documents
Set of 250 ml, 100 ml, and 50 ml beakers with low form design, white graduations, and superior durability and chemical resistance thanks to borosilicate 3.3 glass.
  • Autoclavable, with superior chemical durability
  • Low temperature gradient- can withstand high temperatures and thermal shocks
  • Packaged in a thick, fitted foam casing for safe storage and transport
  • Excellent for science classrooms and professional laboratories
Three piece Beaker set made of high quality, heavy duty, borosilicate 3.3 glass. Includes three, low form beakers with 250 ml, 100 ml and 50 ml capacities. Beakers feature white, printed graduations, beaded rims and spouts for easy pouring. Uniform, thick walls provide excellent strength against breakage. Measurements: (250 ml) 3.8" tall, 2.75" outer diameter, 2.5" inner diameter. 3 mm wall thickness. (100 ml) 2.9" tall, 2" outer diameter, 1.8" inner diameter. 2.5 mm wall thickness. (50 ml) 2.4" tall, 1.75" outer diameter, 1.6" inner diameter. 2 mm wall thickness. Beaker set is packaged in a custom fit, thick foam casing, for safe storage and worry free transport. Microwavable and autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Specifications
  • Capacity:
    50 / 100 / 250 ml
  • Description:
    Premium beaker set