ContactUsLinkComponent
Distilling Glassware Apparatus
The site is currently experiencing technical difficulties. Please retry later. We apologize for any inconvenience.
Distilling Glassware Apparatus
Catalog # 470344-332
Supplier:  Wards
undefined
Distilling Glassware Apparatus
Catalog # 470344-332
Supplier:  Wards
Supplier Number:  CH0890
Specifications
  • Description:
    Distilling glassware apparatus
  • Environmentally Preferable:
  • Cat. no.:
    470344-332
Specifications
About this item
With a 500 ml flask, 300 mm condenser, still head, 100 ml separatory funnel, and an adapter, this distilling glassware apparatus with 19/26 joints ensures efficient and precise distillation processes in various laboratory applications.
  • This distilling kit is made of high quality borosilicate glass
  • The kit consists of only the glass ware for distilling experiments
  • No tubing or heat source or stands are included
  • All joint sizes are 19/26
Everything in the picture is included. Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Contents:
(1) Distilling 500 ml flask
(1) Three way adapter
(1) Liebig condenser with water inlet and outlet tubes
(1) Vacuum adapter
(1) Separatory funnel 100 ml
(1) Flask stopper