Orders LinkContactUsLinkComponent
Sodium Spoon
Sodium Spoon
Sodium spoon
Catalog # 470343-578
Supplier:   Wards
Supplier #:  CH0737
Sodium Spoon
Catalog # 470343-578
Supplier:  Wards
Sodium Spoon
Sodium Spoon
Sodium spoon
Price
$8.60
Log in for availability
Each
Product Details & Documents
The medium-sized sodium spoon is specifically designed for accurate and controlled dispensing of sodium, ensuring safe and efficient handling in experiments.
  • This spoon is for holding sodium under water
  • By compressing handles the sodium spoon opens to allow sodium to be placed in the caged spoon
  • Comprised of rust-proof wire stem and strong cage to safely enclose the sodium
  • Cage measures 15 mm (0.59") diameter
  • Over all Length 248 mm (9.75")
Specifications
  • Description:
    Sodium spoon