Extra Long Crucible Tongs
Catalog # 470343-754
Supplier: Wards
Specifications
- Description:Extra long crucible tongs
- Length:13"
- Cat. no.:470343-754
Specifications
About this item
Designed for furnace use, this extra long 13" stainless steel tong with straight, serrated tips provides a sturdy and reliable grip on crucibles.
- Crucible tongs with extra long handle
- Straight with serrated tips for added grip strength
- Made of high-quality rust- and corrosion-resistant stainless steel
Long and rigid design makes tongs excelled for use in a furnace or other similar equipment. Designed for use in professional laboratories, but is an ideal choice for science classrooms. Tongs measure 13" long, 4" wide when collapsed.
Frequently Bought Together