Funnel and Burette Combined Stand
Catalog #: 470343-590
Supplier: Wards
Specifications
- Description:Funnel and burette combined stand
- Cat. no.:470343-590
Specifications
About this item
Constructed from polished wood, this stand offers a convenient solution for holding both funnels and burettes, ensuring stability during experiments.
- Funnel and burette stand featuring a sturdy hardwood construction that will stand up to the wear and tear of the lab
- Comes with clamping screws
- Designed for use in professional laboratories, but is an ideal choice for science classrooms
This polished wooden funnel and burette stand comes with clamping screws for adjusting the height. Up to 18" height. Base is approximately 11¹/₃" by 4¹/₄".
Customers Who Bought This Also Bought