Stump Base
Stump base
Catalog # 470345-224
Supplier: Wards
Supplier #: CH0656J
Jump to:
Product Details & Documents
Stump base featuring a 2.3" heavy-duty cast iron base in black, providing stability and support for various laboratory equipment and setups.
- Base accommodates 13 mm (0.5") rods (sold separately) secured with a tightening screw
- The durable and sturdy construction of this set makes it an excellent choice for science classroom and for basic professional laboratory setups
Stump retort base, 2.3" Lab stands are used in every area of science - especially chemistry, for holding and manipulating an experimental apparatus. This retort stand is made of chemical-resistant cast iron, and features a hole, suitable for rods with a 13 mm thread. It's base is 2.3" wide and tapers to just over 1.5" at the top with a height of 1.75". Base weighs 1.4 lbs.
Specifications
- Description:Stump base