Mouse Visceral Adipose-specific Serine Protease Inhibitor,Vaspin ELISA Kit
76703-550
:
- Assay Type:Sandwich
- Host:Mouse
- Target protein:Visceral adipose-specific serine protease inhibitor,vaspin
- Description:Mouse Visceral adipose-specific serine protease inhibitor,vaspin ELISA kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Application:Cardiovascular
- Sample volume:50 - 100 μl
- Sensitivity:1 - 5 h
- Regulatory status:RUO
- Cat. no.:76703-550
- Detection wavelength: 450 nm