Mouse Receptor-interacting Serine/Threonine-protein Kinase 3(RIPK3) ELISA Kit
76701-426
:
- Assay Type:Sandwich
- Host:Mouse
- Target protein:Receptor-interacting serine/threonine-protein kinase 3
- Description:Mouse Receptor-interacting serine/threonine-protein kinase 3(RIPK3) ELISA kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Application:Cancer, signal transduction
- Sample volume:50 - 100 μl
- Sensitivity:1 - 5 h
- Regulatory status:RUO
- Cat. no.:76701-426
- Detection wavelength: 450 nm