Mouse Mannan-binding Lectin Serine Protease 2(MASP2) ELISA Kit
Catalog # 76713-098
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:Mannan-binding lectin serine protease 2
- Description:Mouse Mannan-binding lectin serine protease 2(MASP2) ELISA kit
- Size:96 wells
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Storage temperature:2…8 °C
- Sample volume:50 - 100 μl
- Sensitivity:1 - 5 h
- Regulatory status:RUO
- Cat. no.:76713-098
Specifications
About this item
- Detection wavelength: 450 nm